accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNHN

CAS RN 114746-66-6

IUPAC Name: 3-[(piperazin-1-yl)methyl]-1H-indole
Synonyms: 3-(piperazin-1-ylmethyl)-1H-indole
Molecular Weight (g/mol): 215.3
Molecular Formula: C13H17N3
InChi Key: ZTNAIHGOFCMOPJ-UHFFFAOYSA-N
SMILES: C(N1CCNCC1)C1=CNC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results