accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3C

CAS RN 114772-34-8

IUPAC Name: methyl 4'-methyl-[1,1'-biphenyl]-2-carboxylate
Synonyms: methyl 4'-methyl-[1,1'-biphenyl]-2-carboxylate
Molecular Weight (g/mol): 226.28
Molecular Formula: C15H14O2
InChi Key: IHNIAWHITVGYJJ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1C1=CC=C(C)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results