accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 114772-40-6

IUPAC Name: tert-butyl 4'-(bromomethyl)-[1,1'-biphenyl]-2-carboxylate
Synonyms: tert-butyl 4'-(bromomethyl)-[1,1'-biphenyl]-2-carboxylate
Molecular Weight (g/mol): 347.25
Molecular Formula: C18H19BrO2
InChi Key: YHXCWNQNVMAENQ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)C1=CC=CC=C1C1=CC=C(CBr)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results