CAS RN 114812-34-9
IUPAC Name:
1-ethylpyrrolidine-2-carboxamide
Synonyms:
1-ethylpyrrolidine-2-carboxamide
Molecular Weight (g/mol):
142.2
Molecular Formula:
C7H14N2O
InChi Key:
LQOASEHNECNLNM-UHFFFAOYNA-N
SMILES:
CCN1CCCC1C(N)=O
1
–
1
of
1
results
1
(S)-(-)-1-Ethyl-2-pyrrolidinecarboxamide 98.0+%, TCI America™
CAS: 114812-34-9 Molecular Formula: C7H14N2O Molecular Weight (g/mol): 142.202 MDL Number: MFCD00191361 InChI Key: LQOASEHNECNLNM-LURJTMIESA-N Synonym: N-Ethyl-L-prolinamide PubChem CID: 10866443 IUPAC Name: (2S)-1-ethylpyrrolidine-2-carboxamide SMILES: CCN1CCCC1C(=O)N