accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2OOFFFFF

CAS RN 114859-23-3

IUPAC Name: difluoro(2,3,4-trifluorophenyl)methyl 2-methylprop-2-enoate
Synonyms: difluoro(2,3,4-trifluorophenyl)methyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 266.17
Molecular Formula: C11H7F5O2
InChi Key: KYOVZNUXIBOBCQ-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OC(F)(F)C1=C(F)C(F)=C(F)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results