accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONH(S)ClOHO

CAS RN 114873-03-9

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(3-chlorophenyl)propanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-(3-chlorophenyl)propanoic acid
Molecular Weight (g/mol): 299.75
Molecular Formula: C14H18ClNO4
InChi Key: RCZHBTHQISEPPP-NSHDSACASA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC(Cl)=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results