CAS RN 114878-60-3
IUPAC Name:
1-(2-chloro-4-nitrophenyl)piperazine
Synonyms:
1-(2-chloro-4-nitrophenyl)piperazine
Molecular Weight (g/mol):
241.68
Molecular Formula:
C10H12ClN3O2
InChi Key:
JNUWBYPJOBSXDN-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(N2CCNCC2)C(Cl)=C1
1
–
1
of
1
results
1
1-(2-Chloro-4-nitrophenyl)piperazine, 97%, Thermo Scientific™
CAS: 114878-60-3 Molecular Formula: C10H12ClN3O2 Molecular Weight (g/mol): 241.675 MDL Number: MFCD00640776 InChI Key: JNUWBYPJOBSXDN-UHFFFAOYSA-N PubChem CID: 2771331 IUPAC Name: 1-(2-chloro-4-nitrophenyl)piperazine SMILES: C1CN(CCN1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl