accessibility menu, dialog, popup

UserName

Viewing profile as user:
FBrN

CAS RN 114897-91-5

IUPAC Name: 2-(4-bromo-2-fluorophenyl)acetonitrile
Synonyms: 2-(4-bromo-2-fluorophenyl)acetonitrile
Molecular Weight (g/mol): 214.04
Molecular Formula: C8H5BrFN
InChi Key: QLASQEZPJFNZQC-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC=C1CC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results