accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHNNO(E)(E/Z)

CAS RN 1149-16-2

IUPAC Name: 6-{[(1E)-2-[(2-hydroxyphenyl)amino]ethenyl]imino}cyclohexa-2,4-dien-1-one
Synonyms: 6-{[(1E)-2-[(2-hydroxyphenyl)amino]ethenyl]imino}cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 240.26
Molecular Formula: C14H12N2O2
InChi Key: OMJNMRJMNUREOJ-KOBPNRPCSA-N
SMILES: OC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)