accessibility menu, dialog, popup

UserName

Viewing profile as user:
OCaOCuOCuOSrOSrOBiOBiOOBiOBiO

CAS RN 114901-61-0

IUPAC Name: bis([(oxobismuthanyl)oxy]bismuthanone); oxocalcium; bis(oxocopper); bis(oxostrontium)
Synonyms: bis(bismuth(III) oxide); calcium oxide; bis(copper(II) oxide); bis(strontium oxide)
Molecular Weight (g/mol): 1354.32
Molecular Formula: Bi4CaCu2O11Sr2
InChi Key: SCYGCYNQMSDOOE-UHFFFAOYSA-N
SMILES: O=[Ca].O=[Cu].O=[Cu].O=[Sr].O=[Sr].O=[Bi]O[Bi]=O.O=[Bi]O[Bi]=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results