accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 114973-22-7

IUPAC Name: 2,5-difluoro-4-(trifluoromethyl)aniline
Synonyms: 2,5-difluoro-4-(trifluoromethyl)aniline
Molecular Weight (g/mol): 197.11
Molecular Formula: C7H4F5N
InChi Key: FPEXXFHBKBTGJL-UHFFFAOYSA-N
SMILES: NC1=CC(F)=C(C=C1F)C(F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results