accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2

CAS RN 115-07-1

IUPAC Name: prop-1-ene
Synonyms: propylene propene, pure propylene (substance) pure propene propylene preparation
Molecular Weight (g/mol): 42.08
Molecular Formula: C3H6
InChi Key: QQONPFPTGQHPMA-UHFFFAOYSA-N
SMILES: CC=C
Molecular Weight (g/mol) 
  • (2)
  • (1)
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results