accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 115-10-6

H3COCH3

CAS RN 115-10-6

IUPAC Name: methoxymethane
Synonyms: methyl ether 1,1'-oxybis-methane wood ether dymel A methane, 1,1'-oxybis- dimethyl oxide demeon D
Molecular Weight (g/mol): 46.07
Molecular Formula: C2H6O
InChi Key: LCGLNKUTAGEVQW-UHFFFAOYSA-N
SMILES: COC
Molecular Weight (g/mol) 
  • (1)
  • (3)
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)