accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 115-21-9

IUPAC Name: trichloro(ethyl)silane
Synonyms: ethyltrichlorosilane
Molecular Weight (g/mol): 163.5
Molecular Formula: C2H5Cl3Si
InChi Key: ZOYFEXPFPVDYIS-UHFFFAOYSA-N
SMILES: CC[Si](Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results