accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClOOOClClCl

CAS RN 115-27-5

IUPAC Name: 1,7,8,9,10,10-hexachloro-4-oxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Synonyms: chlorendic anhydride
Molecular Weight (g/mol): 370.81
Molecular Formula: C9H2Cl6O3
InChi Key: FLBJFXNAEMSXGL-UHFFFAOYNA-N
SMILES: ClC1=C(Cl)C2(Cl)C3C(C(=O)OC3=O)C1(Cl)C2(Cl)Cl
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)