accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOClClClClCl

CAS RN 115-32-2

IUPAC Name: 2,2,2-trichloro-1,1-bis(4-chlorophenyl)ethan-1-ol
Synonyms: dicofol caswell no. 093 DTMC mitigan keltane para,para'-kelthane kelthane dust base C14H9cl5O kelthane cekudifol
Molecular Weight (g/mol): 370.48
Molecular Formula: C14H9Cl5O
InChi Key: UOAMTSKGCBMZTC-UHFFFAOYSA-N
SMILES: OC(C1=CC=C(Cl)C=C1)(C1=CC=C(Cl)C=C1)C(Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results