CAS RN 115-40-2
Thermo Scientific Chemicals Bromocresol Purple, pure, indicator grade
CAS: 115-40-2 Molecular Formula: C21H16Br2O5S Molecular Weight (g/mol): 540.22 MDL Number: MFCD00011681 InChI Key: ABIUHPWEYMSGSR-UHFFFAOYSA-N Synonym: Bromcresol Purple,BCP,5', 5''-Dibromo-o-cresolsulfonephthalein PubChem CID: 8273 ChEBI: CHEBI:86154 SMILES: CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1
Thermo Scientific Chemicals Bromocresol Purple
CAS: 115-40-2 Molecular Formula: C21H16Br2O5S Molecular Weight (g/mol): 540.22 MDL Number: MFCD00011681 InChI Key: ABIUHPWEYMSGSR-UHFFFAOYSA-N PubChem CID: 8273 ChEBI: CHEBI:86154 IUPAC Name: 2-bromo-4-[3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-6-methylphenol SMILES: CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1
Bromocresol Purple, Sigma-Aldrich
CAS: 115-40-2 | 540.22 g/mol
Bromocresol purple, MP Biomedicals™
CAS: 115-40-2 Molecular Formula: C21H16Br2O5S Molecular Weight (g/mol): 540.22 MDL Number: MFCD00011681 InChI Key: ABIUHPWEYMSGSR-UHFFFAOYSA-N Synonym: 5', 5”-dibromo-o-cresolsulfonephthalein PubChem CID: 8273 ChEBI: CHEBI:86154 IUPAC Name: 3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione SMILES: CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1
Bromocresol Purple (0.04% in Water) [for pH Determination], TCI America™
CAS: 115-40-2 Molecular Formula: C21H16Br2O5S Molecular Weight (g/mol): 540.22 MDL Number: MFCD00011681 InChI Key: ABIUHPWEYMSGSR-UHFFFAOYSA-N PubChem CID: 8273 ChEBI: CHEBI:86154 IUPAC Name: 3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione SMILES: CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1
Bromocresol Purple 97.0+%, TCI America™
CAS: 115-40-2 Molecular Formula: C21H16Br2O5S Molecular Weight (g/mol): 540.22 MDL Number: MFCD00011681 InChI Key: ABIUHPWEYMSGSR-UHFFFAOYSA-N PubChem CID: 8273 ChEBI: CHEBI:86154 IUPAC Name: 3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione SMILES: CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1
Bromocresol Purple, ACS Reagent (Sultone) not water soluble, Reagents
CAS: 115-40-2 Molecular Formula: C21H16Br2O5S Molecular Weight (g/mol): 540.22 InChI Key: ABIUHPWEYMSGSR-UHFFFAOYSA-N Synonym: Bromcresol purple IUPAC Name: 3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione SMILES: CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1
Bromocresol Purple Indicator, Reag. Ph. Eur., Solstice
CAS: 115-40-2 Molecular Formula: C21H16Br2O5S Molecular Weight (g/mol): 540.22 MDL Number: MFCD00011681 InChI Key: ABIUHPWEYMSGSR-UHFFFAOYSA-N PubChem CID: 8273 ChEBI: CHEBI:86154 IUPAC Name: 2-bromo-4-[3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-6-methylphenol SMILES: CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1
Bromocresol Purple (Certified), Fisher Chemical
CAS: 115-40-2 Molecular Formula: C21H16Br2O5S Molecular Weight (g/mol): 540.22 MDL Number: MFCD00011681 InChI Key: ABIUHPWEYMSGSR-UHFFFAOYSA-N Synonym: 5, 5'dibromo-o-cresolsulfonphthalein PubChem CID: 8273 ChEBI: CHEBI:86154 IUPAC Name: 3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione SMILES: CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1