accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 1150-62-5

IUPAC Name: 9-phenyl-9H-carbazole
Synonyms: 9-phenylcarbazole
Molecular Weight (g/mol): 243.31
Molecular Formula: C18H13N
InChi Key: VIJYEGDOKCKUOL-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (3)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)