CAS RN 115029-24-8
IUPAC Name:
2-fluoro-4-(trifluoromethyl)benzoate
Synonyms:
2-fluoro-4-(trifluoromethyl)benzoate
Molecular Weight (g/mol):
207.1
Molecular Formula:
C8H3F4O2
InChi Key:
OCIYTBZXTFPSPI-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC=C(C=C1F)C(F)(F)F
1
–
1
of
1
results
1
Sigma Aldrich 2-Fluoro-4-(trifluoromethyl)benzoic acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.