CAS RN 115055-85-1
IUPAC Name:
zinc(2+) (4-bromophenyl)methanide bromide
Synonyms:
zinc(2+) (4-bromophenyl)methanide bromide
Molecular Weight (g/mol):
315.31
Molecular Formula:
C7H6Br2Zn
InChi Key:
HJQDBIKHGQSYJU-UHFFFAOYSA-M
SMILES:
[Zn++].[Br-].[CH2-]C1=CC=C(Br)C=C1
1
–
1
of
1
results
1
Promotions Available
4-Bromobenzylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
4-Bromobenzylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
CAS: 115055-85-1 Molecular Formula: C7H6Br2Zn Molecular Weight (g/mol): 315.31 MDL Number: MFCD01311435 InChI Key: HJQDBIKHGQSYJU-UHFFFAOYSA-M Synonym: zinc 2+ ion 4-bromophenyl methanide bromide PubChem CID: 57346859 IUPAC Name: zinc(2+) (4-bromophenyl)methanide bromide SMILES: [Zn++].[Br-].[CH2-]C1=CC=C(Br)C=C1