accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1151-94-6

IUPAC Name: (4-methoxyphenyl)(4-nitrophenyl)methanone
Synonyms: (4-methoxyphenyl)(4-nitrophenyl)methanone
Molecular Weight (g/mol): 257.25
Molecular Formula: C14H11NO4
InChi Key: DXVSAELNVPXMSQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results