accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFNNOH2N

CAS RN 1151767-59-7

IUPAC Name: 5-[(3,5-difluorophenyl)methyl]-1,3,4-oxadiazol-2-amine
Synonyms: 5-[(3,5-difluorophenyl)methyl]-1,3,4-oxadiazol-2-amine
Molecular Weight (g/mol): 211.17
Molecular Formula: C9H7F2N3O
InChi Key: RWMWNQXLNITZPU-UHFFFAOYSA-N
SMILES: NC1=NN=C(CC2=CC(F)=CC(F)=C2)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results