accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 115186-31-7

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-6-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-6-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol): 468.55
Molecular Formula: C26H32N2O6
InChi Key: JYEVQYFWINBXJU-ANBDAQEENA-N
SMILES: CC(C)(C)OC(=O)N[C@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results