accessibility menu, dialog, popup

UserName

Viewing profile as user:
OAs

CAS RN 1153-05-5

IUPAC Name: (diphenylarsoroso)benzene
Synonyms: (diphenylarsoroso)benzene
Molecular Weight (g/mol): 322.24
Molecular Formula: C18H15AsO
InChi Key: CLVJBRYGLQNRDA-UHFFFAOYSA-N
SMILES: O=[As](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results