accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 115445-29-9

IUPAC Name: N-ethyl-N-(pyrrolidin-3-yl)acetamide
Synonyms: N-ethyl-N-(pyrrolidin-3-yl)acetamide
Molecular Weight (g/mol): 156.23
Molecular Formula: C8H16N2O
InChi Key: NHMWGGURJSUYGU-UHFFFAOYNA-N
SMILES: CCN(C1CCNC1)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results