accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 115457-83-5

IUPAC Name: [(2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-methoxyoxan-2-yl]methyl N-heptylcarbamate
Synonyms: [(2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-methoxyoxan-2-yl]methyl N-heptylcarbamate
Molecular Weight (g/mol): 335.4
Molecular Formula: C15H29NO7
InChi Key: XPIVOYOQXKNYHA-RGDJUOJXSA-N
SMILES: CCCCCCCNC(=O)OC[C@H]1O[C@H](OC)[C@H](O)[C@@H](O)[C@@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results