accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1154740-65-4

IUPAC Name: 3-chloro-2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: 3-chloro-2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 271.52
Molecular Formula: C12H16BClFNO2
InChi Key: NFNKJOVOJSFUSA-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(Cl)C(F)=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results