accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 115491-93-5

H2COOOOOCH2

CAS RN 115491-93-5

IUPAC Name: bis(prop-2-en-1-yl) dicarbonate
Synonyms: bis(prop-2-en-1-yl) dicarbonate
Molecular Weight (g/mol): 186.16
Molecular Formula: C8H10O5
InChi Key: XVSSGIXTKVRGAR-UHFFFAOYSA-N
SMILES: C=CCOC(=O)OC(=O)OCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)