accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1155-64-2

IUPAC Name: (2S)-2-amino-6-{[(benzyloxy)carbonyl]amino}hexanoic acid
Synonyms: (2S)-2-amino-6-{[(benzyloxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol): 280.32
Molecular Formula: C14H20N2O4
InChi Key: CKGCFBNYQJDIGS-LBPRGKRZSA-N
SMILES: N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)