accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 115520-21-3

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[(triphenylmethyl)sulfanyl]propanoate
Synonyms: 2,3,4,5,6-pentafluorophenyl 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-[(triphenylmethyl)sulfanyl]propanoate
Molecular Weight (g/mol): 751.77
Molecular Formula: C43H30F5NO4S
InChi Key: FDAUCYDVXIPBDR-UHFFFAOYNA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C(CSC(C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(F)=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)