accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CONH2ONH2

CAS RN 115570-52-0

IUPAC Name: 4-[3-(4-aminophenoxy)-2,2-dimethylpropoxy]aniline
Synonyms: 4-[3-(4-aminophenoxy)-2,2-dimethylpropoxy]aniline
Molecular Weight (g/mol): 286.38
Molecular Formula: C17H22N2O2
InChi Key: HPUJEBAZZTZOFL-UHFFFAOYSA-N
SMILES: CC(C)(COC1=CC=C(N)C=C1)COC1=CC=C(N)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results