accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOH

CAS RN 1155843-79-0

IUPAC Name: 2-azatricyclo[3.3.1.1³,⁷]decan-2-ol
Synonyms: 2-azatricyclo[3.3.1.1³,⁷]decan-2-ol
Molecular Weight (g/mol): 153.23
Molecular Formula: C9H15NO
InChi Key: SFXINVPZIZMFHM-UHFFFAOYSA-N
SMILES: ON1C2CC3CC(C2)CC1C3
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results