accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 115661-37-5

IUPAC Name: 2-amino-3-fluorobenzonitrile
Synonyms: 2-amino-3-fluorobenzonitrile
Molecular Weight (g/mol): 136.13
Molecular Formula: C7H5FN2
InChi Key: UNISSOLHERSZOW-UHFFFAOYSA-N
SMILES: NC1=C(C=CC=C1F)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results