accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3NH3CCH3OHO

CAS RN 115662-09-4

IUPAC Name: 6-hydroxy-4,4,10,10-tetramethyl-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene-7-carbaldehyde
Synonyms: 6-hydroxy-4,4,10,10-tetramethyl-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene-7-carbaldehyde
Molecular Weight (g/mol): 273.38
Molecular Formula: C17H23NO2
InChi Key: ZBVWJSQPIHQKQJ-UHFFFAOYSA-N
SMILES: CC1(C)CCN2CCC(C)(C)C3=C(O)C(C=O)=CC1=C23
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results