accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1158830-31-9

CH3SiH3CCH3CH3HO

CAS RN 1158830-31-9

IUPAC Name: diethyl(propan-2-yl)silanol
Synonyms: diethyl(isopropyl)silanol
Molecular Weight (g/mol): 146.31
Molecular Formula: C7H18OSi
InChi Key: MCIZETFFSOLIHL-UHFFFAOYSA-N
SMILES: CC[Si](O)(CC)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results