accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1159-53-1

IUPAC Name: 4-methyl-N,N-bis(4-methylphenyl)aniline
Synonyms: 4-methyl-N,N-bis(4-methylphenyl)aniline
Molecular Weight (g/mol): 287.41
Molecular Formula: C21H21N
InChi Key: YXYUIABODWXVIK-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)