CAS RN 115910-75-3
CAS RN 115910-75-3
IUPAC Name:
N-[1-(propan-2-yloxy)ethyl]acetamide
Synonyms:
N-(1-isopropoxyethyl)acetamide
Molecular Weight (g/mol):
145.2
Molecular Formula:
C7H15NO2
InChi Key:
PUMLTXLDRDSMGQ-UHFFFAOYNA-N
SMILES:
CC(C)OC(C)NC(C)=O
1
–
1
of
1
results
1
N-(1-Isopropoxyethyl)acetamide 93.0+%, TCI America™
CAS: 115910-75-3 Molecular Formula: C7H15NO2 Molecular Weight (g/mol): 145.202 MDL Number: MFCD00191526 InChI Key: PUMLTXLDRDSMGQ-UHFFFAOYSA-N PubChem CID: 14204909 IUPAC Name: N-(1-propan-2-yloxyethyl)acetamide SMILES: CC(C)OC(C)NC(=O)C