accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3MgBr

CAS RN 115913-58-1

IUPAC Name: bromo(4-methylnaphthalen-1-yl)magnesium
Synonyms: bromo(4-methylnaphthalen-1-yl)magnesium
Molecular Weight (g/mol): 245.4
Molecular Formula: C11H9BrMg
InChi Key: PQFCMZXZQYKHIY-UHFFFAOYSA-M
SMILES: CC1=CC=C([Mg]Br)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results