accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 115929-59-4

IUPAC Name: 2-bromo-4-methoxy-6-nitrophenol
Synonyms: 2-bromo-4-methoxy-6-nitrophenol
Molecular Weight (g/mol): 248.03
Molecular Formula: C7H6BrNO4
InChi Key: GAVVLRJHVWIDPK-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=C(O)C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results