accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1159599-99-1

IUPAC Name: 2-oxa-6-azaspiro[3.3]heptane; oxalic acid
Synonyms: 2-oxa-6-azaspiro[3.3]heptane; oxalic acid
Molecular Weight (g/mol): 189.17
Molecular Formula: C7H11NO5
InChi Key: KOUVDKDABFOPIG-UHFFFAOYSA-N
SMILES: OC(=O)C(O)=O.C1NCC11COC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results