accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNOHOHNO

CAS RN 116-25-6

IUPAC Name: 1-(hydroxymethyl)-5,5-dimethylimidazolidine-2,4-dione
Synonyms: mdm hydantoin
Molecular Weight (g/mol): 158.16
Molecular Formula: C6H10N2O3
InChi Key: SIQZJFKTROUNPI-UHFFFAOYSA-N
SMILES: CC1(C)N(CO)C(=O)NC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results