accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 116-29-0

IUPAC Name: 1,2,4-trichloro-5-(4-chlorobenzenesulfonyl)benzene
Synonyms: duphar tetradiphon caswell no. 836 tedane combi tedane tedion tedion V-18 tetradichlone tetrachlorodiphenyl sulfone roztoczol
Molecular Weight (g/mol): 356.04
Molecular Formula: C12H6Cl4O2S
InChi Key: MLGCXEBRWGEOQX-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)S(=O)(=O)C1=CC(Cl)=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results