accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiSSCH3

CAS RN 1160106-12-6

IUPAC Name: 7,7-dioctyl-3,11-dithia-7-silatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraene
Synonyms: 7,7-dioctyl-3,11-dithia-7-silatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraene
Molecular Weight (g/mol): 418.77
Molecular Formula: C24H38S2Si
InChi Key: AUESHXZKMJNIJC-UHFFFAOYSA-N
SMILES: CCCCCCCC[Si]1(CCCCCCCC)C2=C(SC=C2)C2=C1C=CS2
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results