accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1160106-14-8

IUPAC Name: 4,10-dibromo-7,7-dioctyl-3,11-dithia-7-silatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraene
Synonyms: 4,10-dibromo-7,7-dioctyl-3,11-dithia-7-silatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraene
Molecular Weight (g/mol): 576.57
Molecular Formula: C24H36Br2S2Si
InChi Key: PVZQCRDPHHCCRP-UHFFFAOYSA-N
SMILES: CCCCCCCC[Si]1(CCCCCCCC)C2=C(SC(Br)=C2)C2=C1C=C(Br)S2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results