accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 116020-44-1

IUPAC Name: 4-ethenyl-4'-propyl-1,1'-bi(cyclohexane)
Synonyms: 4-ethenyl-4'-propyl-1,1'-bi(cyclohexane)
Molecular Weight (g/mol): 234.43
Molecular Formula: C17H30
InChi Key: KHDBEDDPFRHGCN-UHFFFAOYSA-N
SMILES: CCCC1CCC(CC1)C1CCC(CC1)C=C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results