accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONCH3H3C

CAS RN 116057-81-9

IUPAC Name: N,N-dimethylacetamide
Synonyms: dimethylacetamide hallucinogen N,N'-dimethylacetamide
Molecular Weight (g/mol): 87.12
Molecular Formula: C4H9NO
InChi Key: FXHOOIRPVKKKFG-UHFFFAOYSA-N
SMILES: CN(C)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results