accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 116091-63-5

IUPAC Name: 2-methoxy-5-(2-oxopropyl)benzene-1-sulfonamide
Synonyms: 2-methoxy-5-(2-oxopropyl)benzenesulfonamide
Molecular Weight (g/mol): 243.28
Molecular Formula: C10H13NO4S
InChi Key: MQQJFLHZXQRKKJ-UHFFFAOYSA-N
SMILES: COC1=C(C=C(CC(C)=O)C=C1)S(N)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results