accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 116177-22-1

NNHHOHCl

CAS RN 116177-22-1

IUPAC Name: (1H-imidazol-2-yl)methanol hydrochloride
Synonyms: 1H-imidazol-2-ylmethanol hydrochloride
Molecular Weight (g/mol): 134.56
Molecular Formula: C4H7ClN2O
InChi Key: FEORNCWZOSTSRO-UHFFFAOYSA-N
SMILES: Cl.OCC1=NC=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results