accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNH2

CAS RN 116183-82-5

IUPAC Name: pyrrolidin-3-amine
Synonyms: pyrrolidin-3-amine
Molecular Weight (g/mol): 86.14
Molecular Formula: C4H10N2
InChi Key: NGXSWUFDCSEIOO-UHFFFAOYNA-N
SMILES: NC1CCNC1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results