accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 116247-03-1

IUPAC Name: 2-(pyridin-3-yl)-4,5-dihydro-1,3-thiazole-4-carboxylic acid
Synonyms: 2-(pyridin-3-yl)-4,5-dihydro-1,3-thiazole-4-carboxylic acid
Molecular Weight (g/mol): 208.24
Molecular Formula: C9H8N2O2S
InChi Key: LJGAQGZEJDQDAU-UHFFFAOYNA-N
SMILES: OC(=O)C1CSC(=N1)C1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results